Order Entry
Slovenia
ContactUsLinkComponent
Sodium nitrate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Oxidizing
Irritant
Sodium nitrate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Irritant
Sodium nitrate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • pH (25°C; 5 %):
    5.5 - 8.3
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • IO3 (Iodate):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 20 ppm
  • NO2 (Nitrite):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Fe (Iron):
    Max. 3 ppm
  • Mg (Magnesium):
    Max. 20 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together