Order Entry
Slovenia
ContactUsLinkComponent
Potassium permanganate 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Potassium permanganate 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Potassium permanganate 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Appearance of solution:
    Passes test Ph.Eur.
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Insolubility in water:
    Max. 0.1 %
  • Total N (Nitrogen):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 20 ppm
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together