

Potassium iodate 99.7-100.4%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7758-05-6
:
Formula:KlO₃
MDL Number:MFCD00011406
:214 g/mol
Merck Index:13,07726
Melting Pt:560 °C
Density:3.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-831-9
UN:1479
ADR:5.1,II
:GHS03
:GHS07
Specification Test Results
- Assay:99.7 --> 100.4 %
- pH (20°C; 5 %):5.0 --> 8.0
- Heavy metals (as Pb):Max. 5 ppm
- Insoluble matter:Max. 50 ppm
- Loss on drying (130°C):Max. 0.1 %
- Total N (Nitrogen):Max. 50 ppm
- Cl + ClO3 + Br + BrO3 (as Cl):Max. 100 ppm
- Cl + Br (as Cl):Max. 100 ppm
- I (Iodide):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
- Na (Sodium):Max. 50 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC26840.180 spec_VWRC26840.180 spec_VWRC26840.180 spec_VWRC26840.235 spec_VWRC26840.235
Product specification
More
Frequently Bought Together