
Sodium iodide, ultra dry ≥99.98% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:14,8631
Boiling Pt:1300…1304 °C
Melting Pt:651 °C
Density:3.67
UN:3077
ADR:9,III
:ampouled under argon
EINECS:231-679-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together