Order Entry
Slovenia
ContactUsLinkComponent
Magnetic stirrer heads with ground joint, BOLA
Magnetic stirrer heads with ground joint, BOLA
Supplier:  Bohlender
Certificates
undefined
Magnetic stirrer heads with ground joint, BOLA
Supplier:  Bohlender
Specifications
  • Catalog No:
  • BOHLC450-24
  • BOHLC450-16
  • Speed range:
  • 800 min⁻¹
  • 800 min⁻¹
  • Max. stirring capacity H₂O:
  • 4000 ml
  • 2000 ml