
Potassium carbonate ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:14,7619
Melting Pt:891 °C
Density:2.29
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together