
Tetrachlorocyclopropene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6262-42-6
:
Formula:C₃Cl₄
Boiling Pt:125…130 °C
Density:1,45 g/cm³ (20 °C)
EINECS:228-418-0
ADR:6.1,II
:177.84 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5063 to 1.5083 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:Approx. 0.01 % Citric acid
- Relative density:(20/4°C) 1.026 to 1.036
- Acid value:≤5 mg KOH/g
Specification Test Results
Frequently Bought Together