Menthone (mixture of isomers), Sigma-Aldrich®
Catalog # SIAL63680-100ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:154.25 g/mol
EINECS:233-944-9
CAS Number:10458-14-7
Formula:C₁₀H₁₈O
MDL Number:MFCD00062998