Order Entry
Slovenia
ContactUsLinkComponent
Pesticide reference standard, 2,6-Dichlorobenzamide 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, 2,6-Dichlorobenzamide 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12355000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,6-Dichlorobenzamide 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12355000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    2008-58-4
  • Description:
    2,6-Dichlorobenzamide 10 µg/ml in acetonitrile
  • Formula:
    C₇H₅Cl₂NO
  • Description:
    2,6-Dichlorobenzamide

Specifications

About this item