Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

Flammable
Irritant
   Sustainable Options Available
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Supplier:  Merck
   Sustainable Options Available
Certificates
Cyclohexanone, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SIAL29140-100ML
  • SIAL29140-500ML
  • SIAL29140-1L
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 42,70 €
    42,70 €
    1 * 100 mL
       42,70 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIAL29140-100ML
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 55,70 €
    55,70 €
    1 * 500 mL
       55,70 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIAL29140-500ML
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 98,00 €
    98,00 €
    1 * 1 L
       98,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIAL29140-1L
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.