4-Nitrophenethyl bromide
Glass bottle for solids, 25 g
Catalog # MOLE47797515-25G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
- CAS Number:5339-26-4
- Formula:C₈H₈BrNO₂
- MDL Number:MFCD00007386
- Molecular Weight:230.06 g/mol
- EINECS:226-271-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



