
Cyclopentanone oxime ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1192-28-5
:
Formula:C₅H₉NO
MDL Number:MFCD00001420
:99.13 g/mol
Boiling Pt:196 °C
Melting Pt:54…58 °C
Flash Pt:92 °C (198 °F)
Storage Temperature:Ambient
EINECS:214-749-8
:GHS06
Frequently Bought Together