



Myrcene 90% tech. stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-35-3
:
Formula:C₁₀H₁₆
MDL Number:MFCD00008908
:136.24 g/mol
Boiling Pt:165 °C (760 mmHg)
Density:0.795 g/cm³
Flash Pt:44 °C
EINECS:204-622-5
UN:2319
ADR:3,III
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4660 to 1.4720 (20°C, 589 nm)
- Density:(20/4°C) 0.789 to 0.800 g/ml
- GC:≥88 %
- Stabilizer:≤0.15 % alpha-Tocopherol
Specification Test Results
Frequently Bought Together