3-(Trimethylsilyl)-2-propyn-1-ol, Sigma-Aldrich®
Catalog # SIAL318558-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:128.25 g/mol
EINECS:226-094-5
UN:2810
CAS Number:5272-36-6
Formula:C₆H₁₂OSi
MDL Number:MFCD00002913