Order Entry
Slovenia
ContactUsLinkComponent
3-Bromo-3-buten-1-ol 97+%
Irritant
3-Bromo-3-buten-1-ol 97+%
Certificates
undefined
Irritant
3-Bromo-3-buten-1-ol 97+%

Specification Test Results

  • Appearance:
    Clear brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5000 to 1.5020 (20°C, 589 nm)
  • GC:
    ≥97.0 %

Specification Test Results

Related Information

Frequently Bought Together