Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Acetyl-5-bromo-7-nitroindoline
1-Acetyl-5-bromo-7-nitroindoline, Sigma-Aldrich®
1-Acetyl-5-bromo-7-nitroindoline, Sigma-Aldrich®
SIALA6250-1G
:
Merck
:
1-Acetyl-5-bromo-7-nitroindoline, Sigma-Aldrich®
SIALA6250-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
285.1 g/mol
:
62368-07-4
:
C₁₀H₉BrN₂O₃
:
MFCD00056018