2-Methyl-1-hexene, Sigma-Aldrich®
SIAL111627-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:98.19 g/mol
:92 °C (1013 hPa)
:–103 °C
:0.697 g/cm³ (20 °C)
:Ambient
:228-041-1
:2287
:3,II
:M-Clarity™ quality level = MQ100
:6094-02-6
:C₇H₁₄
:MFCD00009486