Order Entry
Slovenia
ContactUsLinkComponent
3-Methyl-1-butyne 96%
Irritant
Flammable
3-Methyl-1-butyne 96%
Certificates
undefined
Irritant
Flammable
3-Methyl-1-butyne 96%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3730 to 1.3750 (20°C, 589 nm)
  • GC:
    ≥95.0 %

Specification Test Results

Related Information

Frequently Bought Together