5-Iodovanillin 97%
: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde
:5438-36-8
:
:C₈H₇IO₃
:MFCD00006941
:335
:278.05 g/mol
:180…184 °C
:226-617-7
:GHS07
- Environmentally Preferable:
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:180 to 184 °C
- HPLC:≥96.0 %
- UV:at 250 nm A: ≤0.05
Frequently Bought Together