
4-Fluoro-3-nitrobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:453-71-4
:
Formula:C₇H₄FNO₄
MDL Number:MFCD00007058
:185.11 g/mol
Boiling Pt:92 °C (15 mmHg)
Melting Pt:121…126 °C
Storage Temperature:Ambient
EINECS:207-221-3
:GHS07
Related Information
Frequently Bought Together