2-Chloro-4-nitrobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-60-5
:
Formula:C₇H₄ClNO₄
MDL Number:MFCD00007209
Molecular Weight:201.57 g/mol
Melting Pt:139…143 °C
EINECS:202-771-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:138 to 144 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together