
3-Hydroxy-4-nitrobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:619-14-7
:
Formula:C₇H₅NO₅
MDL Number:MFCD00007110
:183.12 g/mol
Melting Pt:233 °C
EINECS:210-580-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:≥228 °C
- Water:≤0.5 %
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together