4-Aminobenzonitrile 98%
:873-74-5
:
:C₇H₆N₂
:MFCD00007821
:118.14 g/mol
:167 °C (1,1 -rr)
:83…87 °C
:1,274 g/cm³ (20 °C)
:Ambient
:212-850-1
:3439
:6.1,III
:GHS06
- Appearance:Light yellow to beige to orange to grey-brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:83 to 87 °C
- GC:≥97.5 %
Frequently Bought Together