Order Entry
Slovenia
ContactUsLinkComponent
Organic reference standard, m-Cresol 1,000 µg/ml in methanol
Organic reference standard, m-Cresol 1,000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, m-Cresol 1,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    108-39-4
  • Formula:
    C₇H₈O
  • Description:
    m-Cresol

Specifications

Frequently Bought Together