Organic reference standard, 1,1,2-Trichlorotrifluoroethane 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL17788300ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:76-13-1
- Description:1,1,2-Trichlorotrifluoroethane 10 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₂Cl₃F₃
- Description:1,1,2-Trichlorotrifluoroethane