Order Entry
Slovenia
ContactUsLinkComponent
Heptadecafluorooctanesulphonyl fluoride
Heptadecafluorooctanesulphonyl fluoride
Catalog # EHERC15987130
Supplier:  EHRENSTORFER
CAS Number:  
Heptadecafluorooctanesulphonyl fluoride
Catalog # EHERC15987130
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Perfluorooctanesulphonyl fluoride
 :
 :502.12 g/mol
EINECS:206-200-6
CAS Number:307-35-7
Formula:C₈F₁₈O₂S
MDL Number:MFCD00012016