Magnesium citrate hydrate (∼ 16% Mg) (dried basis) (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3344-18-1
:
Formula:C₁₂H₁₀Mg₃O₁₄·xH₂O
MDL Number:MFCD09953657
:451.13 g/mol (anhy)
Merck Index:14,5663
Storage Temperature:Ambient
EINECS:222-093-9
Related Information
Frequently Bought Together