
1-Bromo-4-chloro-2-nitrobenzene
Supplier: CHEMPUR
Certificates
About this item
CAS Number:41513-04-6
:
Formula:C₆H₃BrClNO₂
MDL Number:MFCD00024320
:236.45 g/mol
Melting Pt:70 °C
Density:2,048 g/cm³ (20 °C)
EINECS:255-421-4
:GHS07
Frequently Bought Together