1-Bromo-4-chlorobenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-39-8
:
Formula:C₆H₄BrCl
MDL Number:MFCD00000600
:191.45 g/mol
Boiling Pt:196 °C (760 mmHg)
Melting Pt:64…67 °C
Density:1.651 g/cm³
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:203-392-3
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:64 to 67 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together