Isopropenyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-22-5
:
Formula:C₅H₈O₂
MDL Number:MFCD00008709
:100.12 g/mol
Merck Index:13,05223
Boiling Pt:94 °C (1013 hPa)
Melting Pt:–93 °C
Density:0.9188 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-562-7
UN:2403
ADR:3,II
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together