Filtration capsules, Sartopore® 2
Ready to use disposable capsules for sterile filtration in laboratory scale or for small scale filterability trials.
- Ideal for scale-up and scale-down
- Compatible with a pH range from pH one to 14
- Typical applications include sterilising grade filtration of therapeutics, biological fluids, injectables, purified water, media, buffers
Hydrophilic PES membrane, PP housing.
Each individual element is tested for integrity by bubble point and diffusion test prior to be released assuring absolute reliability.
- Catalog No:
- SARP5445307H9-FF-A
- SARP5445307H7-SO-A
- SARP5445307H8-SO-A
- SARP5445307HS-HH-M
- SARP5445307H0-FF-V
- SARP5441307H4G-OOB
- SARP5445307H8G-OOA
- SARP5445306G7-OO-A
- SARP5445307H9-SO-A
- SARP5445307H7-SS-A
- SARP5445358K7-SO-A
- SARP5445307G7-FF-A
- SARP5445307H7-FF-A
- Type:
- Sartopore® 2 MidiCaps size 9
- Sartopore® 2 MidiCaps size 7
- Sartopore® 2 MidiCaps size 8
- Sartopore® 2 SartoScale
- Sartopore® 2 MidiCaps size 0
- Sartopore® 2 size 4
- Sartopore® 2 Gamma MidiCaps size 8
- Sartopore® 2 MidiCaps size 7
- Sartopore® 2 MidiCaps size 9
- Sartopore® 2 MidiCaps size 7
- Sartopore® 2 MidiCaps size 7
- Sartopore® 2 MidiCaps size 7
- Sartopore® 2 MidiCaps size 7
- Inlet/outlet:
- Tri-clamp 25 mm (³/₄)/tri-clamp 25 mm (³/₄")
- SO (1¹/₂" sanitary flange/hose barb)
- Tri-clamp 25 mm (1¹/₂")/¹/₂" single stepped hose barb
- ¹/₄'' multiple stepped hose barb / ¹/₄'' multiple stepped hose barb
- Tri-clamp (³/₄)/tri-clamp (³/₄")
- ¹/₄'' multiple stepped hose barb
- 1" single stepped hose barb/1" single stepped hose barb
- ¹/₂ single stepped hose barb/¹/₂" single stepped hose barb
- Tri-clamp 25 mm (1¹/₂")/¹/₂" single stepped hose barb
- SS (1¹/₂" sanitary flange / 1¹/₂" sanitary flange)
- SO (1¹/₂" sanitary flange/hose barb)
- ³/"₄ tri-clamp 25 mm/³/₄" tri-clamp 25mm
- Tri-clamp 25 mm (³/₄)/tri-clamp 25 mm (³/₄")
- Ø×L:
- 77×197 mm
- 77×115 mm
- 77×148 mm
- 77×330 mm
- 77×154 mm
- 77×115 mm
- 77×197 mm
- 77×115 mm
- 77×115 mm
- 77×115 mm
- 77×115 mm
- Max. pressure:
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 4 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
- 5 bar @ 20 °C; 2 bar @ 80 °C
Frequently Bought Together