2,4-Dimethyl-3-pentanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,4-Dimethylpentan-3-one, Diisopropyl ketone, Isobutyrone
CAS Number:565-80-0
:
Formula:C₇H₁₄O
MDL Number:MFCD00008918
:114.19 g/mol
Boiling Pt:124 °C (1013 hPa)
Melting Pt:–80 °C
Density:0.8 g/cm³
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:209-294-7
UN:1224
ADR:3,II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4020 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together