2-Ethylhexanal ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 123-05-7 Molecular Formula: C8H16O Formula Weight: 128.21 Flash Point: 50°(122°F) Refractive Index: 1.4155 Melting Point: -100° Boiling Point: 163° Beilstein Reference: 1700556 UN No.: 1989 MDL No.: MFCD00006987
- CAS Number:123-05-7
- Formula:C₈H₁₆O
- MDL Number:MFCD00006987
- Molecular Weight:128.21 g/mol
- Boiling Pt:163 °C
- Melting Pt:< –100 °C
- Density:0.819
- Flash Pt:50 °C (122 °F)
- Storage Temperature:Ambient
- EINECS:204-596-5
- UN:1989
- ADR:3,III
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 123-05-7 Molecular Formula: C8H16O Formula Weight: 128.21 Flash Point: 50°(122°F) Refractive Index: 1.4155 Melting Point: -100° Boiling Point: 163° Beilstein Reference: 1700556 UN No.: 1989 MDL No.: MFCD00006987
- CAS Number:123-05-7
- Formula:C₈H₁₆O
- MDL Number:MFCD00006987
- Molecular Weight:128.21 g/mol
- Boiling Pt:163 °C
- Melting Pt:< –100 °C
- Density:0.819
- Flash Pt:50 °C (122 °F)
- Storage Temperature:Ambient
- EINECS:204-596-5
- UN:1989
- ADR:3,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



