2-Ethylhexanal 96%
- :123-05-7
- :C₈H₁₆O
- :MFCD00006987
- :128.21 g/mol
- :163 °C (1013 hPa)
- :–90 °C
- :0.8195 g/cm³
- :50 °C
- :Ambient
- :204-596-5
- :1191
- :3,III
- :123-05-7
- :C₈H₁₆O
- :MFCD00006987
- :128.21 g/mol
- :163 °C (1013 hPa)
- :–90 °C
- :0.8195 g/cm³
- :50 °C
- :Ambient
- :204-596-5
- :1191
- :3,III
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4180 (20°C, 589 nm)
- GC:≥95 %
- Specific gravity:(20/20°C) 0.819 to 0.824
- Acid value:≤8 mg KOH/g
Product Family Options
- Pack typePk
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



