ω-Bromo-2-hydroxyacetophenone, Sigma-Aldrich®
Catalog # SIAL594938-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.05
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:3263
CAS Number:2491-36-3
Formula:C₈H₇BrO₂
MDL Number:MFCD01727570