Order Entry
Slovenia
ContactUsLinkComponent
 
 
 
2,2,2-Trichloroethanol 99%
 
undefined
 
 
2,2,2-Trichloroethanol 99%
 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4880 to 1.4910 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together