1-Chlorobutane, Sequenation Grade
ACRO404550010
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- GC:≥99.5 %
- UV:at 220 nm A: ≤1
- Residue after evaporation:≤2 ppm
: n-Butyl Chloride
:
:92.57 g/mol
:13,01559
:77…78 °C (760 mmHg)
:–123 °C
:0.88 g/cm³
:–12 °C
:Ambient
:203-696-6
:1127
:3,II
:GHS02
:GHS08
:109-69-3
:CH₃(CH₂)₃Cl
:MFCD00001009