Bismuth triacetate ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22306-37-2
:
Formula:(H₃CCOO)₃Bi
MDL Number:MFCD00015629
:386.11 g/mol
Melting Pt: Dec.
Storage Temperature:Ambient
EINECS:244-904-5
Related Information
Frequently Bought Together