Pesticide reference standard, 2,3,4,5,6-Pentachloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15940000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:527-20-8
- Description:2,3,4,5,6-Pentachloroaniline 10 µg/ml in cyclohexane
- Formula:C₆H₂Cl₅N
- Description:2,3,4,5,6-Pentachloroaniline