Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

7-Nitroindole

Glass bottle for solids, 25 g
Catalog # COBBIN-0019-25G
Supplier:   COMBI-BLOCKS
7-Nitroindole
Catalog # COBBIN-0019-25G
Supplier:  COMBI-BLOCKS
CAS Number:  6960-42-5
7-Nitroindole

Glass bottle for solids, 25 g

CAS Number:  6960-42-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:6960-42-5
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD00005683
  • Molecular Weight:162.15 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications