
Methyl-4-iodo-3-nitrobenzoate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89976-27-2
:
Formula:C₈H₆INO₄
MDL Number:MFCD08064223
:307.04 g/mol
Melting Pt:101…104 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together