Order Entry
Slovenia
ContactUsLinkComponent
3-Furonitrile, Sigma-Aldrich®
3-Furonitrile, Sigma-Aldrich®
Catalog # SIAL715700-1G
Supplier:  Merck
CAS Number:  
3-Furonitrile, Sigma-Aldrich®
Catalog # SIAL715700-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :93.08 g/mol
Boiling Pt:151 °C (1013 hPa)
Melting Pt:37…38 °C
UN:2926
ADR:4.1,III
CAS Number:30078-65-0
Formula:C₅H₃NO
MDL Number:MFCD00067042