Order Entry
Slovenia
ContactUsLinkComponent
3-Amino-9-ethyl carbazole (AEC)
Health
3-Amino-9-ethyl carbazole (AEC)
Catalog # MOLE22225310-100G
Supplier:  Molekula
CAS Number:  
Health
3-Amino-9-ethyl carbazole (AEC)
Catalog # MOLE22225310-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :210.28 g/mol
Melting Pt:113…114 °C
Storage Temperature:Ambient
EINECS:205-057-7
UN:2811
ADR:6.1,III
 :GHS08
CAS Number:132-32-1
Formula:C₁₄H₁₄N₂
MDL Number:MFCD00004964