Order Entry
Slovenia
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)-1H-pyrazole
3,5-Bis(trifluoromethyl)-1H-pyrazole
Catalog # MAYBAC31181.10
Supplier:  Thermo Scientific
CAS Number:  
3,5-Bis(trifluoromethyl)-1H-pyrazole
Catalog # MAYBAC31181.10
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Boiling Pt:148 °C (1013 hPa)
 :204.07 g/mol
CAS Number:14704-41-7
Formula:C₅H₂F₆N₂
MDL Number:MFCD00153672
Boiling Pt:148 °C (1013 hPa)