2-Furonitrile
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:617-90-3
:
Formula:C₅H₃NO
MDL Number:MFCD00003223
:93.08 g/mol
Boiling Pt:146 °C (1013 hPa)
Density:1.085 g/cm³ (20 °C)
EINECS:210-537-4
UN:1992
ADR:3,III
Frequently Bought Together