


3-Chloro-5-fluoroaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4863-91-6
:
Formula:C₆H₅ClFN
MDL Number:MFCD03407962
ADR:6.1,III
:145.56 g/mol
EINECS:None
UN:2810
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together