Vacuum filtration manifold, Nalgene®
Catalog # 513-3451
Supplier: Thermo Fisher Scientific
Specifications
- Description:Vacuum filter funnel manifold, stainless steel, outlet: 9,5 mm (0,38 in.)
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:1
Specifications
About this item
Vacuum filter funnel manifold for use with Nalgene analytical filter funnels and test filter funnels.
- Made from stainless steel
- Two-way valve with vent port