D-(+)-Camphor
ROTH6155.1
:
- Pack type:Glass bottle for solids
- Pk:250 g
:
:152.24 g/mol
:13,01739
:408 °C (1013 hPa)
:174…175 °C
:0.992 g/cm³ (20 °C)
:66 °C
:Ambient
:207-355-2
:2717
:4.1,III
:GHS02
:GHS07
:464-49-3
:C₁₀H₁₆O
:MFCD00064149