Order Entry
Slovenia
ContactUsLinkComponent
D-(+)-Camphor
Flammable
Irritant
D-(+)-Camphor
Catalog # MOLEM49280091
Supplier:  Molekula
CAS Number:  
Flammable
Irritant
D-(+)-Camphor
Catalog # MOLEM49280091
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.24 g/mol
Merck Index:13,01739
Boiling Pt:408 °C (1013 hPa)
Melting Pt:174…175 °C
Density:0.992 g/cm³ (20 °C)
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:207-355-2
UN:2717
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:464-49-3
Formula:C₁₀H₁₆O
MDL Number:MFCD00064149