Order Entry
Slovenia
ContactUsLinkComponent
D-(+)-Camphor 97%
D-(+)-Camphor 97%
Certificates
undefined
D-(+)-Camphor 97%

Specification Test Results

  • Appearance:
    White Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    174 to 179 °C
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +41° to +44° (20 °C, 589 nm) (c=10, alcohol)

Specification Test Results

Related Information

Frequently Bought Together